Advantec Sanitary Stainless Steel Pressure Filtration Holders
Stainless steel type 304 or 316 minimizes resistance and maximizes flow rate
Pressure filtration holders maximize flow rates by applying positive pressure to the fluid over a broad membrane surface. Holders are autoclavable with one filter membrane that is sealed with removable wing nuts. All holders vent to release pressure.
Holders have PTFE gaskets, silicone O-rings, and PTFE-coated photoetched stainless steel support screens to prevent nitrocellulose membranes from sticking.
Sanitary holders are best used in-line to filter-sterilize liquids or gases. Sanitary inlet and outlet connections can be completely disassembled for thorough cleaning. Top plate has knobs for easy positioning over membrane.
$15,634.14CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Filter Diameter (mm) Material Prefilter Diameter (mm)
Availability Pricing
Advantec 302800 316 SS Pressure Filtration Holder for 293 mm Membranes RK-29900-10 Mfr # 302800
COMPARE
293 316 Stainless Steel 257
$15,634.14CAD / Each