Dynalon Adjustable Volume Dispenser Bottles and Measuring Cup
Easily adjust dispensing volume
  • Suitable for foodstuff
The adjustable volume LDPE bottle features a graduated clear PMP measuring cup that slides vertically on the polyethylene tube for easy adjustments. To dispense, set the volume to be dispensed and gently squeeze the bottle to fill the cup. When the bottle pressure is released, the excess will be drawn back into the bottle; only the desired volume will remain.
$390.45 - $419.92CAD / Case of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Max Graduation (mL)
Availability Pricing
Dynalon Adjustable-Volume Dispenser Bottle, LDPE/PMP, 250mL; 10/CS RK-07879-50 Mfr # 206305
COMPARE
250 Low Density Polyethyene (LDPE) 25
$390.45CAD / Case of 10
Dynalon Adjustable-Volume Dispenser Bottle, LDPE/PMP, 500mL; 10/CS RK-07879-52 Mfr # 206315
COMPARE
500 Low Density Polyethyene (LDPE) 25
$419.92CAD / Case of 10