Adjustable Plexiglas Splash Shields
Fully adjustable for maximum flexibility
Shields are constructed of 1/8" Plexiglas® to protect from spash or flying debris. Shields move side to side as well as up and down - maximizing valuable workspace when not in use. Flexible arms adjust to maximum 38" depth. Use brackets (must order separately) to mount shields directly to edge of counter to secure in place. Use convenient side handles to position shield angle for maximum protection.
$1,165.45CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Height (in) Width (in)
Availability Pricing
Scienceware F24966-0003 Adjustable Plexiglas Splash Shield, 15" W x 24" H RK-86469-02 Mfr # F24966-0003
COMPARE
24 12
$1,165.45CAD / Each