This product is now discontinued, View Related Products
Adam Equipment Ultrasonic Height and Digital Health Scale (MUW-300L)
Fast results and simple operation for use in locker rooms, offices, homes and more
  • Robust metal housing
  • Aluminum pillar
  • Anti-slip platform
  • Base has five built-in feet for added stability
  • Height measurements up to 6.8 ft (210 cm)
  • Height measurements are taken using ultrasonic technology
  • Output data to a computer or printer through the RS-232 interface
  • Large, sealed backlit display
  • Rechargeable battery and AC adapter included
  • Automated BMI function for body mass index reading
  • Other features include overload protection, tare range, hold function, and security locking point
$1,466.02 - $1,510.22CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment MUW-250L Ultrasonic Height and Digital Health Scale, 600lb/300kg 115V RK-11120-76 Mfr # MUW-300L-115V
COMPARE
300 660 115
$1,466.02CAD / Each
Adam Equipment MUW-300L Ultrasonic Height and Digital Health Scale, 600lb/300kg 230V RK-11120-77 Mfr # MUW-300L-230V
COMPARE
300 660 230
$1,510.22CAD / Each