Adam Equipment PTM Low Profile Industrial Platform Scales with Display
The PTM low profile platform is ideal for drums and wheelchairs.
  • Adjustable feet
  • Non-slip adjustable levelling feet
  • Four load cell construction
  • Checked plate for added grip
  • Check weighing LED lights provided on either indicator - GK or AE402
  • Approximately 10 foot long shielded cable
  • Wheels to allow easy movement
  • Available with a choice of indicators
  • Built in ramps on each side for easy access
  • Flat weighing surface excludes ramps: 580 x 760mm (including ramps 800 x 932mm)
The easy access ramps on both sides offers smooth transitions from the floor to the weighing platform and the diamond checked platform offers a durable weighing area. With handles and wheels. Available with GK indicator or IP66 rated AE402 indicator.
$2,335.32CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment PTM 500 & GK Indicator, 1100lb/500kg Capacity, 0.2Lb/100g Readability, Universal Voltage RK-11810-46 Mfr # PTM 500 [Gk]
COMPARE
500 1100 115 / 220
$2,335.32CAD / Each
Adam Equipment PTM 500 & GK Indicator, 1100lb/500kg Capacity, 0.2Lb/100g Readability, 220V RK-11810-47 Mfr # PTM 500 [G] 220V
COMPARE
500 1100 230
$2,335.32CAD / Each