This product is now discontinued, View Related Products
Adam Equipment Nimbus Toploading Balances
Reliable performance in a compact, rugged design
  • All-metal housing ensures rugged, durable protection and stability
  • GLP compliant printouts via RS-232 connection
  • Weighing applications include parts counting, check weighing, percent weighing, dynamic/animal weighing
  • Below-balance weighing hook
  • Date and time stored in memory
  • Large backlit display and intuitive keypad
Optimized connections boost data collection capabilities and lead to exceptional information transmission. USB and RS-232 interfaces are standard, while an additional interface allows use of an optional remote display. Whether performing basic tasks such as printing data, or advanced communication with a LIMS system, the Nimbus can meet the requirements. GLP printouts are available with time, date and other essential information.
Innovative design creates a smaller footprint, so the Nimbus occupies minimal space on the lab bench. Solid metal construction throughout the balance durably withstands chemicals and rigorous daily use. Optimized configuration of the Nimbus’s internal weighing system results in enhanced performance. Fabricated from a single block, the weighing sensor contains fewer parts than a traditional force motor balance, improving efficiency.
$1,024.01 - $2,408.99CAD / Each
44 variations of this product are available.
See All Product Options Below
44 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Adam Equipment NBL 223-115V Nimbus Precision Toploader Balance, 220g x 1mg, Backlit Lcd, 115V RK-11121-06 Mfr # NBL 223E-115V
COMPARE
220 115 0.001
$1,318.68CAD / Each
Adam Equipment NBL 223-230V Nimbus Precision Toploader Balance, 220g x 1mg, Backlit Lcd, 230V RK-11121-07 Mfr # NBL 223E-230V
COMPARE
220 230 0.001
$1,318.68CAD / Each
Adam Equipment NBL 423-115V Nimbus Precision Toploader Balance, 420g x 1mg, Backlit Lcd, 115V RK-11121-08 Mfr # NBL 423E-115V
COMPARE
420 115 0.001
$1,554.43CAD / Each
Adam Equipment NBL 423-230V Nimbus Precision Toploader Balance, 420g x 1mg, Backlit Lcd, 230V RK-11121-09 Mfr # NBL 423E-230V
COMPARE
420 230 0.001
$1,554.43CAD / Each
Adam Equipment NBL 2602-115V Nimbus Precision Toploader Balance, 2600g x 10mg, Backlit Lcd, 115V RK-11121-11 Mfr # NBL 2602E-115V
COMPARE
2600 115 0.01
$1,318.68CAD / Each
Adam Equipment NBL 2602-230V Nimbus Precision Toploader Balance, 2600g x 10mg Backlit Lcd, 230V RK-11121-12 Mfr # NBL 2602E-230V
COMPARE
2600 230 0.01
$1,318.68CAD / Each
Adam Equipment NBL 3602-115V Nimbus Precision Toploader Balance, 3600g x 10mg, Backlit Lcd, 115V RK-11121-13 Mfr # NBL 3602E-115V
COMPARE
3600 115 0.01
$1,466.02CAD / Each
Adam Equipment NBL 3602-230V Nimbus Precision Toploader Balance, 3600g x 10mg, Backlit Lcd, 230V RK-11121-14 Mfr # NBL 3602E-230V
COMPARE
3600 230 0.01
$1,466.02CAD / Each
Adam Equipment NBL 4602-115V Nimbus Precision Toploader Balance, 4600g x 10mg, Backlit Lcd, 115V RK-11121-16 Mfr # NBL 4602E-115V
COMPARE
4600 115 0.01
$1,613.36CAD / Each
Adam Equipment NBL 4602-230V Nimbus Precision Toploader Balance, 4600g x 10mg, Backlit Lcd, 230V RK-11121-17 Mfr # NBL 4602E-230V
COMPARE
4600 230 0.01
$1,613.36CAD / Each
Adam Equipment NBL 3201-115V Nimbus Precision Toploader Balance, 3200G x 0.1g, Backlit Lcd, 115V RK-11121-18 Mfr # NBL 3201-115V
COMPARE
3200 115 0.1
$1,024.01CAD / Each
Adam Equipment NBL 3201-230V Nimbus Precision Toploader Balance, 3200G x 0.1g, Backlit Lcd, 230V RK-11121-19 Mfr # NBL 3201-230V
COMPARE
3200 230 0.1
$1,024.01CAD / Each
Adam Equipment NBL 4201-115V Nimbus Precision Toploader Balance, 4200g x 1.0g, Backlit Lcd, 115V RK-11121-20 Mfr # NBL 4201E-115V
COMPARE
4200 115 0.1
$1,245.01CAD / Each
Adam Equipment NBL 4201-230V Nimbus Precision Toploader Balance, 4200g x 1.0g, Backlit Lcd, 230V RK-11121-21 Mfr # NBL 4201E-230V
COMPARE
4200 230 0.1
$1,245.01CAD / Each
Adam Equipment NBL 8201-115V Nimbus Precision Toploader Balance, 8200g x 1.0g, Backlit Lcd, 115V RK-11121-22 Mfr # NBL 8201E-115V
COMPARE
8200 115 0.1
$1,510.22CAD / Each
Adam Equipment NBL 8201-230V Nimbus Precision Toploader Balance, 8200g x 1.0g, Backlit Lcd, 230V RK-11121-23 Mfr # NBL 8201E-230V
COMPARE
8200 230 0.1
$1,510.22CAD / Each
Adam Equipment Nimbus NBL 1602i Precision Toploading Balance, 1600 g x 0.01 g with Internal Cal, 115 V RK-11122-48 Mfr # NBL 1602I 115V
COMPARE
1600 115 0.01
$1,510.22CAD / Each
Adam Equipment Nimbus NBL 1602i Precision Toploading Balance, 1600 g x 0.01 g with Internal Ca, 230 V RK-11122-49 Mfr # NBL 1602I 230V
COMPARE
1600 230 0.01
$1,510.22CAD / Each
Adam Equipment Nimbus NBL 223i Precision Toploading Balance, 220 g x 1 mg with Internal Cal, 115 V RK-11122-58 Mfr # NBL 223I 115V
COMPARE
220 115 0.001
$1,583.89CAD / Each
Adam Equipment Nimbus NBL 223i Precision Toploading Balance, 220 g x 1 mg with Internal Cal, 230 V RK-11122-59 Mfr # NBL 223I 230V
COMPARE
220 230 0.001
$1,583.89CAD / Each
Showing 1 - 20 of 44