Adam Equipment Mechanical Health Scales
Fast results and simple operation for use in locker rooms, offices, homes, and more
  • Robust metal housing
  • Aluminum pillar
  • Anti-slip platform
  • Base with five built-in feet for added stability
  • Height measurements up to 6.8 ft (210 cm)
  • 100% mechanical operation that requires no external power supply or batteries
  • Built-in wheels allow easy movement
  • Adjustable zero point
MDW health and fitness scales offer an affordable and durable solution for medical facilities, and provide practical features for school locker rooms and fitness centers. All MDW scales feature an adjustable height rod for precise height readings. The MDW 160M is a mechanical scale with wheels and a notched counter weight that clearly shows the weight.
$434.65 - $581.99CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Description
Availability Pricing
Adam Equipment MDW 200B  Mechanical Health Scale, 440 lb/200 kg x 0.2 lb/0.1 kg RK-11119-31 Mfr # MDW 200B
COMPARE
200 440 Mechanical Health Scale, 440 lb/200 kg x 0.2 lb/0.1 kg
$434.65CAD / Each
Adam Equipment MDW 200M  Mechanical Health Scale, 440 lb/200 kg x 0.2 lb/0.1 kg with Height Rod & Wheels RK-11119-32 Mfr # MDW 200M
COMPARE
200 440 Mechanical Health Scale, 440 lb/200 kg x 0.2 lb/0.1 kg with Height Rod & Wheels
$581.99CAD / Each