Adam Equipment GFK Industrial Scales
Built tough to meet a variety of applications
  • Battery and AC operation
  • Full numeric keypad lets you preset tare, select sample size (count mode), and lock out functions to prevent unauthorized tampering with scale settings
  • Tilt or rotate the adjustable pillar-mounted indicator to suit your viewing needs
  • Large backlit LCD with capacity tracker helps prevent overload
  • Parts counting with selectable sample size
  • Checkweighing LED lights show acceptable limits
  • Removable stainless steel pan for easy cleaning
  • Automatic accumulation for adding totals
  • Bidirectional RS-232 output
Built tough for a variety of industries the GFK can meet the demands of the warehouse, or the production floor. Check weighing applications are trouble free with amber, green, and red indicators to show the operator if the item is under, over, or at the preset limits.
$1,495.49 - $2,121.68CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment GFK 1320A Industrial Scale, 1320lb/600kg Capacity and 0.1Lb/50g Readability, 230V RK-11124-00 Mfr # GFK 1320A
COMPARE
600 1320 230
$2,040.65CAD / Each
Adam Equipment GFK 165A Industrial Scale, 165B/75kg Capacity and 0.002Lb/1g Readability, 230V RK-11124-02 Mfr # GFK 165A
COMPARE
75 165 230
$1,495.49CAD / Each
Adam Equipment GFK 660A Industrial Scale, 660lb/300kg and 0.05Lb/20g Readability, 230V RK-11124-05 Mfr # GFK 300
COMPARE
300 230
$1,554.43CAD / Each
Adam Equipment GFK 165A Industrial Scale, 165Lb/75kg and 0.01Lb/5G Readability RK-11710-00 Mfr # GFK 165A
COMPARE
75 165 115
$1,495.49CAD / Each
Adam Equipment GFK 165AH Floor Checkweighing Scale, 165lb/75kg x 0.002lb/1g RK-11710-01 Mfr # GFK 165AH
COMPARE
75 165 100 to 240
$2,062.75CAD / Each
Adam Equipment GFK 330A Floor Checkweighing Scale, 330lb/150kg x 0.02lb/10g RK-11710-02 Mfr # GFK 330A
COMPARE
150 330 100 to 240
$1,495.49CAD / Each
Adam Equipment GFK 330AH Floor Checkweighing Scale, 330lb/150kg x 0.005lb/2g RK-11710-03 Mfr # GFK 330AH
COMPARE
150 330 115
$2,062.75CAD / Each
Adam Equipment GFK 660A Floor Checkweighing Scale, 660lb/300kg x 0.05lb/20g RK-11710-04 Mfr # GFK 660A
COMPARE
300 660 100 to 240
$1,613.36CAD / Each
Adam Equipment GFK 1320A Floor Checkweighing Scale, 1320lb/600kg x 0.1lb/50g RK-11710-05 Mfr # GFK 1320A
COMPARE
600 1320 115
$2,121.68CAD / Each
Showing 1- 9 of 9