Adam Equipment Aqua ABW-S Stainless Steel Washdown Scales
IP68 rated— designed to withstand harsh working environments
  • Sealed membrane keypad protects against dirt and spills
  • 304 SS casing and pan offer easy cleaning and durability
  • Weighing modes include parts counting, percentage weighing, and check weighing
  • Overload protection prevents damage to internal components
  • Dual LED displays—located in the front and rear of balance
  • Fast stabilization time of 1 second
  • Internal rechargeable battery for operation anywhere
The Aqua stainless steel washdown scales with IP68 rating are ideal for use in messy and dusty environments. The 304 stainless steel housing with removable pan and sealed keypad can be hosed down for a quick clean up. The bright dual LED displays located in the front and rear of the scale allow multiple users to use the same scale.
Check weighing is fast and simple with built-in LED indicators that clearly tell you if the load is under, over, or within acceptable limits. The parts counting optimization feature automatically refines piece weight as parts are added. External calibration feature ensures accurate measurements. Use the balance almost anywhere—a rechargeable battery lasts up to 55 hours of operation. Auto power-off conserves battery life. Heavy-duty rubber feet ensure stability of unit.
$795.63CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Readability (g)
Availability Pricing
Adam Equipment ABW 16S Aqua Stainless Steel Washdown Scale, 32lb / 16kg x 0.002lb / 1g RK-55001-14 Mfr # ABW 16S
COMPARE
16 32 1
$795.63CAD / Each
Adam Equipment ABW 4S Aqua Stainless Steel Washdown Scale, 9lb / 4kg x 0.0002lb / 0.1g RK-55001-15 Mfr # ABW 4S
COMPARE
4 9 0.1
$795.63CAD / Each
Adam Equipment ABW 32S Aqua Stainless Steel Washdown Scale, 70lb / 32kg x 0.004lb / 2g RK-55001-16 Mfr # ABW 32S
COMPARE
32 70 2
$795.63CAD / Each
Adam Equipment ABW 8S Aqua Stainless Steel Washdown Scale, 18lb / 8kg x 0.001lb / 0.5g RK-55001-17 Mfr # ABW 8S
COMPARE
8 18 0.5
$795.63CAD / Each