This product is now discontinued, View Related Products
Adam Equipment AE 402 Industrial Scale Indicator
Wash down meets IP66 standards
  • Large backlit LCD
  • Color coded keys
  • Rechargeable battery
  • Bi-directional RS-232 interface
  • Bracket for wall mounting
  • Hold function
  • Stainless steel housing
  • Checkweighing LED lights
  • Capacity tracker
The AE402 indicator is a stainless steel indicator that has a range of features for many different weighing applications. Whether these indicators are used outdoors, on a loading dock or in a warehouse or industrial buildings the stainless steel housing is sealed for IP66 wash down environments and the quality construction is built tough to handle even the most demanding tasks. The sleek compact design comes complete with the wall mount bracket and hardware to place it where you want it. A built-in rechargeable battery with low battery indicator, check weighing limit LED, an easy-to-use keypad with highlighted tare key and a large backlit display complete the AE402 indicator.
$383.08CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
Adam Equipment AE 402 Stainless Steel Washdown Scale Indicator, IP66; 115V RK-11710-56 Mfr # AE 402
COMPARE
115 Stainless Steel Ip66 Washdown Scale Indicator
$383.08CAD / Each
Adam Equipment AE 402 (220V) Stainless Steel Washdown Scale Indicator, IP66; 220V RK-11710-57 Mfr # AE 402 (220V)
COMPARE
230 Stainless Steel Ip66 Washdown Scale Indicator 220V
$383.08CAD / Each