Cole-Parmer® Essentials Acrylic Burettes (Burets)
No leaking stopcock
  • Durable burette (buret) is constructed with a clear, acrylic body
Clear acrylic burette (buret) features leakproof stopcock with self lubricating PTFE plug. Burette is individually calibrated for maximum accuracy and is resistant to dilute mineral acids and bases with the exception of HF and ammonium hydroxide. Can be used in solutions with pH ranging from 1 to 14, and with up to 5% W/W of acids or 30% W/W of bases.
$94.74 - $151.76CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Description
Availability Pricing
COMPARE
25 0.1 Acrylic Burette, 25 mL; 1/Pk
$94.74CAD / Each
COMPARE
50 0.1 Acrylic Burette, 50 mL; 1/Pk
$100.93CAD / Each
COMPARE
100 0.1 Acrylic Burette, 100 mL; 1/Pk
$151.76CAD / Each