This product is now discontinued, View Related Products
Accuform Danger - Use Lockout Before Working on Equiprment Signs
Protect your workers by clearly displaying possible hazards
These signs indicate potential, type, and degree of the hazard that may lead to accidental injury or property damage. Signs define specific hazards by conveying a clear, easily understood message. The sign headers, and colors used in accident prevention safety signs, are specified by OSHA and ANSI to designate the severity of the hazard. Each color has a specific role.
Danger (red) indicates an imminently hazardous situation which, if not avoided, will result in death or serious injury. Lockout/Tagout (red) advises of a lockout/tagout situation; if not avoided, will result in death or serious injury.
Signs are available in adhesive vinyl, plastic, and aluminum to best suit your application. Adhesive vinyl is a flexible 4-mil pressure-sensitive vinyl with adhesive back and peel-off liner. This material is ideal for low-cost, light-duty applications. Plastic is 0.055" polyethylene plastic and is good for medium duty indoor environments and sheltered outdoor environments. Aluminum is 0.040" thick with overlaminate coat for long-term outdoor exposure. It withstands splashes and washdown without rusting.
$16.94 - $44.20CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Width (in)
Availability Pricing
Accuform MLKT021VS Safety Sign, Danger Lockout Equipment 7 X 10, Adhesive Vinyl RK-40711-09 Mfr # MLKT021VS
COMPARE
7 Adhesive Vinyl 10
$16.94CAD / Each
Accuform MLKT021VP Safety Sign, Lockout Equipment Using Lockout, 7 X 10, Plastic RK-40711-10 Mfr # MLKT021VP
COMPARE
7 Plastic 10
$18.42CAD / Each
Accuform MLKT021VA Safety Sign, Danger Lockout Equipment Using Lockout, 7 X 10, Aluminum RK-40711-11 Mfr # MLKT021VA
COMPARE
7 Aluminum 10
$26.52CAD / Each
Accuform MLKT016VS Safety Sign, Danger Use Lockout Before Working On Equipment, 10x14 Adhesive Vinyl RK-40711-29 Mfr # MLKT016VS
COMPARE
10 Adhesive Vinyl 14
$25.78CAD / Each
Accuform MLKT016VP Safety Sign, Danger Use Lockout Before Working On Equipment, 10x14 Plastic RK-40711-30 Mfr # MLKT016VP
COMPARE
10 Plastic 14
$27.99CAD / Each
Accuform MLKT016VA Safety Sign, Danger Use Lockout Before Working On Equipment, 10x14 Aluminum RK-40711-31 Mfr # MLKT016VA
COMPARE
10 Aluminum 14
$44.20CAD / Each