A&D Weighing Ion Semi and Micro Analytical Balances
Weigh trace quantities with confidence – even fine powders
  • Built-in fan-less ionizer eliminates static interference
  • Interlocking doors allow you to open either door from opposite side
  • Auto-calibration when balance detects a charge in ambient temperature
  • Automatic response adjustment reduces effects from draft and vibration
  • Built-in sensors monitor, temperature, humidity, vibration and atmospheric pressure
  • Be compliant -- external data logger stores up to 5000 weighing results
These balances use a fan-less, integrated, direct current ionizer to neutralize charged samples. A green LED lights up when the ionizer is on. Transparent metal coating on the glass further minimizes the effects of static.
A bight backlit LCD allows you to easily read measurements. The units also feature one-touch digital internal calibration with auto self-cal (ASC), dual draft rings to reduce draft interferences, and a removable separation plate for weighing taller objects.
$14,262.42 - $28,222.79CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
A&D Weighing BM-20 Ion Micro Balance, 22 g x 0.001 mg RK-11110-89 Mfr # BM-20
COMPARE
Internal 22 0.001
$28,222.79CAD / Each
A&D Weighing BM-22 Ion Micro Balance, 5.1/22 g x 0.001/0.01 mg RK-11110-90 Mfr # BM-22
COMPARE
Internal 22 0.001
$26,793.60CAD / Each
A&D Weighing BM-252 Ion Semi-Micro Balance, 250 g x 0.01 mg RK-11110-91 Mfr # BM-252
COMPARE
Internal 250 0.01
$14,262.42CAD / Each
A&D Weighing BM-5 Ion Micro Balance, 5.2 g x 0.001 mg RK-11113-50 Mfr # BM-5
COMPARE
Internal 5.2 0.001
$25,747.49CAD / Each
A&D Weighing BM-5D Ion Micro Balance, 2.1/5.2 g x 0.001/0.01 mg RK-11113-51 Mfr # BM-5D
COMPARE
Internal 5.2 0.001
$24,318.30CAD / Each