This product is now discontinued, View Related Products
400 Series Thermistor Probes
Probes are accurate to +/- 0.1 deg C from 0 to 70 deg C. Electrically isolated probes come with a nondetachable 10-ft lead with phone plug (except as noted).
Water-resistant junctions between probe and lead should not be immersed; detachable leads are not water resistant. Note: Time constant is estimate time required to reach 63.2% of a new reading. To determine how long it will take to reach 99% of a new reading multiply the time constant by five.
1/4" or 6.350 mm phono plug connection
$19.15 - $284.36CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Connector Type Max Temperature (° F) Sheath Length (in)
Availability Pricing
COMPARE
Phono plug 212 0.3125
$198.91CAD / Each
COMPARE
Phono plug 212 13
$181.23CAD / Each
COMPARE
Phono plug 302 4.5
$251.95CAD / Each
COMPARE
Phono plug 302 4.5
$269.63CAD / Each
COMPARE
300 1.875
$282.89CAD / Each
COMPARE
300 0.40625
$228.74CAD / Each
Measurement Specialties 409B 400 Series Thermistor Attachable Surface Probe RK-08442-15 Mfr # 409B
COMPARE
Phono plug 212 0.375
$203.33CAD / Each
COMPARE
300 0.1875
$250.48CAD / Each
COMPARE
Phono plug 300 4.5
$269.63CAD / Each
COMPARE
302 5
$250.48CAD / Each
Measurement Specialties 44033 400 Series Thermistor Probes; Epoxy-Encapsulated Thermistor Element RK-08459-10 Mfr # 44033
COMPARE
167 0.1875
$19.15CAD / Each
COMPARE
300 4.6875
$223.96CAD / Each
COMPARE
Phono plug 212 2.5
$284.36CAD / Each
Showing 1- 13 of 13