This product is now discontinued, View Related Products
3M Air Mate Belt Mounted (PAPR)
High efficiency filters for extremely fine dust hazards
  • Up to eight hours operation on a single charge
This positive pressure hooded respirator is ideal for workers with facial hair who can not wear tight fitting mask. It is an excellent choice in applications where there is very fine dust, such as in asbestos abatement. Assembled components comprise a NIOSH approved respirator. Order components separately.
The standard hood features a chin shroud with elastic band that helps maintain a tight face fit. The extended bib hood can be tucked into workers coveralls allowing a flow of air to help cool the user. Both style hoods are made from Tychem® for excellent particulate and liquid protection.
The breathing tube brings air from blower assembly to the hood.
Blower assembly includes blower, HEPA filter with 99.97% efficiency to 0.3 micron, battery (charger sold separately), waist belt, and airflow indicator.
$111.98 - $2,073.80CAD / Pkg of 3
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
3M BE-12-3 Belt-Mounted PAPR Respirator Hood, 3/Pk RK-40220-11 Mfr # BE-12-3
COMPARE
Belt-Mounted PAPR Respirator Hood, 3/Pk
$277.00CAD / Pkg of 3
3M BE-10-3 Respirator Hood, Extended Length Pkg/3 RK-40220-12 Mfr # BE-10-3
COMPARE
Respirator Hood, Extended Length Pkg/3
$257.84CAD / Pkg of 3
3M Air-Mate Respirator Breathing Tube RK-40220-13 Mfr # BE-224
COMPARE
Respirator Breathing Tube
$111.98CAD / Each
3M 231-01-30 Powered Air Purifying Respirator Blower Assy RK-40220-14 Mfr # 231-01-30
COMPARE
Powered Air Purifying Respirator Blower Assy
$2,073.80CAD / Each
3M Air-Mate HEPA Respirator Filter RK-40220-15 Mfr # 451-02-01R01
COMPARE
HEPA Respirator Filter
$131.13CAD / Each
3M 520-03-73 Respirator Blower Battery Charger RK-40220-16 Mfr # 520-03-73
COMPARE
Respirator Blower Battery Charger
$479.59CAD / Each