This product is now discontinued, View Related Products
3M 7000 Series Silicone Respirators
Light silicone full facepiece enhances fit and improves durability
  • Wide, scratch-resistant, polycarbonate face shield provides excellent visibility and protection
  • Extra-large inhalation and exhalation valves for easier breathing
Respirator includes a a standard nosecup to prevent fogging and a scratch resistant polycarbonate lens for greater durability. A high quality, rust-free speaking diaphragm facilitates excellent communication. A double flap face seal helps ensure proper fit and protection and four adjustable straps help distribute the respirators weight for better comfort.
Order respirator cartridges separately according to the demands of your work environment. Versatile respirator accepts 6000 series bayonet cartridges.
Approval: NIOSH
$810.36 - $833.94CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Size
Availability Pricing
3M 7000 Series Silicone Respirator, Small RK-86459-00 Mfr # 7800S
COMPARE
Small
$810.36CAD / Each
3M 7000 Series Silicone Respirator, Mediuml RK-86459-02 Mfr # 7800M
COMPARE
Medium
$833.94CAD / Each
3M 7000 Series Silicone Respirator, Large RK-86459-04 Mfr # 7800S-L
COMPARE
Large
$833.94CAD / Each