This product is now discontinued, View Related Products
1.5-mm Combs for Cole-Parmer Horizontal Mid-Size Gel Systems
Combs are single sided and height adjustable; available in three thicknesses to accommodate different sample volumes. The 13- and 27- well combs are spaced to allow sample loading with multi-channel pipettors.
$92.46 - $97.98CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Comb for Horizontal Mid-Size Gel System; 10 Wells, 1.5 mm Thick
$92.46CAD / Each
COMPARE
Comb for Horizontal Mid-Size Gel System; 13 Wells, 1.5 mm Thick
$92.46CAD / Each
COMPARE
Comb for Horizontal Mid-Size Gel System; 16 Wells, 1.5 mm Thick
$92.46CAD / Each
COMPARE
Comb for Horizontal Mid-Size Gel System; 20 Wells, 1.5 mm Thick
$95.03CAD / Each
COMPARE
Comb for Horizontal Mid-Size Gel System; 27 Wells, 1.5 mm Thick
$97.98CAD / Each