This item is now discontinued. View the direct replacement: RK-29700-86 or browse similar items
ZenPure 50mm in-line disc filter, Polypropylene, 5.0um, 10/pk
Representative image only.
ZenPure Mfr # D50CP0502H2H-10 – Item # RK-49700-86
Simple, quick, and efficient filtration of liquids and gases
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolypropylene
  • Housing MaterialPolypropylene
  • Pore Size (µm)5
  • Filter Diameter (mm)50
  • Effective Filter Area (cm²)15.9
  • Connections1/4\" to 3/8\" stepped barb
  • Max Pressure (PSI)60
  • Description50mm in-line disc filter, Polypropylene, 5.0um, 10/pk
More About this Item
Available in a large variety of membrane materials and pore sizes for filtration of liquids and gases. The compact design of the filter capsules reduces hold-up volume and exposure to hazardous chemicals. Use these filter devices in-line or connected to a syringe. Housing is made of polypropylene for excellent chemical compatibility. No adhesives, binders, or surfactants are used in the manufacturing process.