This item is now discontinued. Please browse similar items
YSI 900 Chlorine Free and Total Chlorine Colorimeter Kit
Representative image only.
YSI Mfr # 250900Y – Item # RK-99550-20
Single-parameter colorimeter for quick and accurate measurements
  • US EPA-approved DPD method
  • Large, easy-to-read backlit LCD
  • Durable, IP68-rated waterproof housing
  • Convenient data storage with GLP time-and-date stamp
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Display TypeBacklit LCD
  • Light SourceLED
  • Wavelength(s) (nm)530
  • Test ParametersChlorine; free and total
  • Data LoggingYes
  • Memory Size16 data sets
  • Battery4 x AAA batteries (included)
  • Length (in)6 1/2
  • Width (in)3
  • Height (in)1 1/2
  • DescriptionFree and Total Chlorine Colorimeter Kit
  • Warranty2 year
Two year warrantyConformite EuropeenneEPA United States Environmental protection agencyIP68 Rated
Key Features
  • US EPA-approved DPD method
  • Large, easy-to-read backlit LCD
  • Durable, IP68-rated waterproof housing
  • Convenient data storage with GLP time-and-date stamp
  • 2 year warranty
More About this Item
The YSI 900 rugged, waterproof chlorine colorimeter is ideal for laboratory, field, and plant measurements. Innovative light shield is designed to avoid breakage or loss. The meter features a large, backlit display, a real-time clock, and automatic data logging for 16 data sets. User calibration is available to any government or organizational standards (NIST, ISO, or DIN).