DWK Life Sciences (Wheaton) W900180-6 Histology Columbia Staining Jar w/Cover, Glass; 6/Cs
Representative image only.
DWK Life Sciences (Wheaton) Mfr # W900180-6 – Item # RK-48585-52
Staining Dish for Cover Slip
$447.17CAD / Case of 6
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHistology Columbia Staining Jar w/Cover, Glass; 6/Cs
More About this Item
This staining jar holds up to four cover slips, measuring 17 to 23 mm wide and up to 30 mm long. Longer cover slips can be accommodated if cap is removed. Glass meets USP Type III standards and
resists most chemicals. Jar includes a white polypropylene screw cap with PTFE coated polyethylene liner. Also use this jar for cleaning cover slips, as well as coating them with materials such as polylysine
or silane.