DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 5-10 Slides

Representative image only.
DWK Life Sciences (Wheaton) Mfr # 900470 – Item # RK-48585-30
Ideal for staining slides
  • Constructed of durable soda-lime glass
  • Jar is designed to hold 3” x 1” (75mm x 25mm) slides
  • Jar holds 5 single 3” x 1” (75mm x 25mm) slides vertically or 10 slides back-to-back

PRODUCT OPTIONS

$102.40CAD / Each
Include Slides Today...
Cole-Parmer® Essentials Microscope Slides
$403.71 - $610.35CAD / Case of 1440

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Number of slides)5 single or 10 back-to-back
  • Internal Width (in)1
  • Internal Height (in)3 19/32
  • Internal Depth (in)1 1/10
  • Internal Width (cm)2.6
  • Internal Height (cm)9.2
  • Internal Depth (cm)2.8
  • DescriptionCoplin Staining Jar with Cover, Glass, 5-10 Slides

Key Features

  • Constructed of durable soda-lime glass
  • Jar is designed to hold 3” x 1” (75mm x 25mm) slides
  • Jar holds 5 single 3” x 1” (75mm x 25mm) slides vertically or 10 slides back-to-back

More About this Item

WHEATON® Coplin staining jars provide a stable container for staining slides. Glass covers protect slides from outside elements and reduce reagent evaporation. Compact size makes it easy to transfer slides and requires only a small volume of reagent.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat