This item is now discontinued. Please browse similar items
DWK Life Sciences (Wheaton) 358204 7-Piece Micro Tissue Grinder Kit with Case
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 358204 – Item # RK-04468-48
Complete selection of micro tissue grinders for small samples
  • Polyethylene case with foam inserts provides protection
  • Tissue grinder with screw cap (0.5 mL only) is compatible with Wheaton Connection® components
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Description7-Piece Micro Tissue Grinder Kit with Case
  • Warranty1 year
1 Year Warranty
Key Features
  • Polyethylene case with foam inserts provides protection
  • Tissue grinder with screw cap (0.5 mL only) is compatible with Wheaton Connection® components
  • 1 year warranty
More About this Item
Kit contains 1 mL Tenbroeck, 0.5 mL tissue grinder with screw cap, 1 mL dounce, 2 mL Potter-Elvehjem, 1 mL tapered, 0.1 mL Potter-Elvehjem, and 0.2 mL micro tissue grinders.