Whatman 1541-150 541 Ashless Quantitative Filter Paper, 20-25um, 15cm; 100/Box
Representative image only.
Whatman Mfr # 1541-150 – Item # RK-06647-35
The industry standard for high-purity filtration
  • Available in a variety of grades and sizes
PRODUCT OPTIONS
$136.29CAD / of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCellulose
  • Pore Size (micron)23
  • Diameter (mm)150
  • Diameter (cm)15
  • Description541 Ashless Quantitative Filter Paper, 20-25um, 15cm; 100/Box
Key Features
  • Available in a variety of grades and sizes
More About this Item
Our Whatman® filter papers offer you flexibility in filter choice without sacrificing quality. All filters are packaged 100 per box.
Quantitative filter papers are the choice for gravimetric analysis. High-purity hardened filters have a high wet-strength and are chemical-resistant to handle vacuum filtration or acid/alkali solutions.