Whatman 1005-125 Qualitative Filter Papers; 12.5 cm dia; pore size, 2.5 µ; 100/box
Representative image only.
Whatman Mfr # 1005-125 – Item # RK-06648-24
The industry standard for high-purity filtration
  • Available in a variety of grades and sizes
  • 45 day warranty
PRODUCT OPTIONS
$65.42CAD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • Pore Size (micron)2.5
  • Diameter (mm)125
  • Diameter (cm)12.5
  • DescriptionQualitative Filter Papers; 12.5 cm dia; pore size, 2.5 µ; 100/box
  • Warranty45 days
Key Features
  • Available in a variety of grades and sizes
  • 45 day warranty
More About this Item
Our Whatman® filter papers offer you flexibility in filter choice without sacrificing quality. All filters are packaged 100 per box.
Qualitative filter papers are used for liquid clarifications, analytical separations, or air and water analysis.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.