This item is now discontinued. Please browse similar items
Weller 037103169372 Professional butane soldering pen kit
Representative image only.
Weller Mfr # P2KC – Item # RK-97105-61
Soldering pen equivalent to a 25 to 75 watt soldering iron
  • ESD safe to protect sensitive components
  • Pen has an automatic cut-off switch when the cap is replaced
SPECS & DESCRIPTION
Specifications & Description
  • Set IncludesP2C soldering tool, PPT9 hot air tip, PPT10 hot knife tip, PPT11 hot air deflector, PPT12 flame tip
  • DescriptionProfessional butane soldering pen kit
Key Features
  • ESD safe to protect sensitive components
  • Pen has an automatic cut-off switch when the cap is replaced
More About this Item
This lightweight soldering pen features professional grade quality in a compact design. Operate as a soldering iron or a heat tool. Gas catalyst system produces predictable and repeatable temperatures to ensure professional results. Convenient push-button switch offers quick and convenient ignition. Torch is powered by butane; order separately below.