This item is now discontinued. Please browse similar items
Waterless ice bucket kit; two chill packs, bead bag - NO BEADS
Representative image only.
Item # RK-06274-28
Avoid contamination risk – dry beads provide ice-free cooling
  • Several hours of constant cooling
  • Durable and lightweight — polyurethane bucket with molded side knobs
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionWaterless ice bucket kit; two chill packs, bead bag - NO BEADS
Key Features
  • Several hours of constant cooling
  • Durable and lightweight — polyurethane bucket with molded side knobs
More About this Item
Keep your samples cold, clean, and dry. This ice-free bucket helps you avoid water puddles and float-away tubes. Use anywhere in place of traditional ice. Simply cool the beads and chill packs in your refrigerator or freezer, then assemble the chill bucket. Put the chill packs on the bottom, and place filled bead bag in the bucket. Create your own temperature range by starting with beads at temperatures at ambient, or cooled to 4°C, -20°C, or -80°C. May substitute chill packs with dry ice.
What's included: bucket, lid, two reusable chill packs, and nylon bag.