VistaLab Ovation M M Mechanical Volume Starter Set Pipette Kit, 2, 20, 200, 1000 uL
Representative image only.
VistaLab Mfr # 1070-0999 – Item # RK-06199-20
Ergonomics and performance with innovation you can feel
  • Easily contours to your hand for ultimate comfort
  • Only minimal force is required in the proven ergonomic design
  • Built-in cap opener at base opens PCR and microtubes with ease
  • 1 year warranty
PRODUCT OPTIONS
$1,801.96CAD / Each
Often Bought With…
VistaLab™ VistaRak™ Pipette Tips
$103.14 - $324.15CAD / Case of 960
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)1000
  • Min Volume (µL)0.2
  • DescriptionM Mechanical Volume Starter Set Pipette Kit, 2, 20, 200, 1000 uL
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards AssociationETL ListedISO9001 Registered SupplierUL Listed.
Key Features
  • Easily contours to your hand for ultimate comfort
  • Only minimal force is required in the proven ergonomic design
  • Built-in cap opener at base opens PCR and microtubes with ease
  • 1 year warranty
More About this Item
With value in mind, these mechanical adjustable-volume pipettors use the same proven ergonomic Ovation design to minimize contact pressure, tension, and exertion.
The low force plunger allows you to manually control speed with a light touch of the thumb. Releasing pipette tips is a breeze with just a quick press of the tip eject button. The one-way nozzle protects your pipettor and samples as it blocks access of liquid into the body of the pipettor.
For maximum efficiency, use the built-in cap opener at the base of the pipettor to open tops of PCR and microtubes. No pipette stands are necessary, the Ovation pipettor stands by itself on the benchtop. Order the optional stand to hold up to four pipettors at a time, saving you valuable bench space in the lab.
Ovation M Starter Set includes (2µL, 20µL, 200µL, 1000µL)