This item is now discontinued. Please browse similar items
Vector Laboratories DuoLux Chemiluminescent/Fluorescent Substrate for Peroxidase, 200 ml
Representative image only.
Vector Laboratories Mfr # SK-6604-200 – Item # RK-93952-08
Blots luminesce and fluoresce for easy visualization
  • Strong luminescent signal lasts up to 8 hours
  • Fluorescence is permanent and detectable for months
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ColorChemiluminescent, fluorescent
  • Volume (mL)200
  • ConjugatePeroxidase
  • DescriptionChemiluminescent/Fluorescent Substrate for Peroxidase, 200 ml
  • Warranty1 year
1 Year Warranty
Key Features
  • Strong luminescent signal lasts up to 8 hours
  • Fluorescence is permanent and detectable for months
  • 1 year warranty
More About this Item
DuoLux substrate kits are available for both peroxidase and alkaline phosphatase reactions to produce a strong chemiluminescent and fluorescent signal. The substrate luminesces in the blue range, with a peak emission at 453 nm. Fluorescent excitation occurs over a range of wavelengths: 254 nm, 365 nm, with maximum excitation at 405 nm. Maximum fluorescent emission occurs at 453 nm.