This item is now discontinued. Please browse similar items
BrandTech 636657 VSK3000 Vacuum Gauge for 07379-06
Representative image only.
BrandTech Mfr # 20636657 – Item # RK-07379-91
Gauge head for your Vaccubrand vacuum controller
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVSK3000 Vacuum Gauge for 07379-06
More About this Item
Designed for use with Vacuubrand vacuum gauges and controllers 636657, 674291, 683160 and 683170. This is the gauge head only. This is external pressure transducer with a capacitive, ceramic diaphragm sensor with 1080 to 0.1 mbar range.