This item is now discontinued. Please browse similar items
Uvex by Honeywell S4200HS Protégé Safety Glasses, Metallic Black Frame, Clear HydroShield Anti-Fog Lens; 10/Bx
Representative image only.
Uvex by Honeywell Mfr # S4200HS – Item # RK-86497-88
Ensure safety with enhanced anti-fog technology and greater scratch resistance
  • Advanced HydroShield® anti-fog design requires no application or maintenance Secure wrap around frame extends to fit a wide range of users Cushioned nosepiece and temple pads minimize slippage and provides a comfortable fit
  • Designed with metallic black frame color
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorMetallic Black
  • Lens ColorClear
  • Lens CoatingHydroShield® anti-fog
  • Lens TypePolycarbonate
  • ComplianceANSI Z87.1-2010 and CSA Z94.3
  • DescriptionProtégé Safety Glasses, Metallic Black Frame, Clear HydroShield Anti-Fog Lens; 10/Bx
Canadian Standards Association
Key Features
  • Advanced HydroShield® anti-fog design requires no application or maintenance Secure wrap around frame extends to fit a wide range of users Cushioned nosepiece and temple pads minimize slippage and provides a comfortable fit
  • Designed with metallic black frame color
More About this Item
Created with innovative Floating Lens™ design, these safety glasses provide workers with exceptional comfort and flexibility. Weighing less than one ounce, the ultra-lightweight construction offers an all-day comfortable fit.
Clear polycarbonate lenses feature HydroShield anti-fog technology, keeping lenses consistently fog-free for up to 90 times longer. Avoid fogging when temperature or humidity changes, reducing down time and exposure to injury.
Compliance: ANSI Z87.1-2010 and CSA Z94.3 standard