This item is now discontinued. Please browse similar items
Uvex by Honeywell S2960HS Hypershock Safety Glasses, Brown Frame, Clear, HydroShield Antifog Lens; 10/BX
Representative image only.
Uvex by Honeywell Mfr # S2960HS – Item # RK-86497-97
High-performance wraparound eyewear with a sporty style
  • Protects against impact from sun, wind and dust, as well as airborne debris HydroShield long-life antifog coating to help keep lenses clear Soft molded inserts and nosepiece provide a comfortable, secure fit
  • Brown Frame
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorBrown
  • Lens ColorClear
  • Lens CoatingAntifog
  • ComplianceANSI Z87.1-2015, CSA Z94.3
  • DescriptionSafety Glasses, Brown Frame, Clear, Hydroshield Antifog Lens; 10/BX
Key Features
  • Protects against impact from sun, wind and dust, as well as airborne debris HydroShield long-life antifog coating to help keep lenses clear Soft molded inserts and nosepiece provide a comfortable, secure fit
  • Brown Frame
More About this Item
Sharp and modern looking, these clear safety glasses have superior side protection and peripheral vision. Hypershock glasses are 100-percent dielectric so, you can wear them in environments where accidental exposure to an electrical charge is possible. Polycarbonate lens features antiscratch and HydroShield antifog hard coatings.
Compliance: ANSI Z87.1-2015, CSA Z94.3