This item is now discontinued. Please browse similar items
United Electric J120-188 Switch 50/1000psi 316Lss Diaphragm Explosion-Proof Pressure Switch
Representative image only.
United Electric Mfr # J120-188 – Item # RK-68456-96
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (PSI)50
  • Max Pressure (PSI)1000
  • Wetted Materials316L SS, Viton® GLT
  • Process Connection1/2\" NPT(F)
  • Electrical Connections3/4\" conduit
  • Electrical terminationScrew terminals
  • Switch output15 A at 125/250 VAC (resistive), NO or NC
  • OutputSPDT
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionSwitch 50/1000psi 316Lss Diaphragm Explosion-Proof Pressure Switch
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Legendary UE reliability
  • Explosion-proof
  • Class I, Div 1 & 2, (Zone 1); Class II, Div 1 & 2; Class III
  • Worldwide approvals and certifications
  • Terminal block wiring
  • Dual conduits
  • Enclosure design permits safe and easy access for wiring
  • Internal adjustment
  • Die cast aluminum enclosure, epoxy powder coated and gasketed
  • REVIEWS