This item is now discontinued. Please browse similar items
United Electric E121-5BS Explosion-Proof Temperature Switch, -20/80F; Remote Mount
Representative image only.
United Electric Mfr # E121-5BS – Item # RK-39008-29
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • RelaysSPDT
  • Min Temperature (° F)-20
  • Max Temperature (° F)80
  • DescriptionExplosion-Proof Temperature Switch, -20/80F; Remote Mount
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Legendary UE reliability
  • Explosion-proof
  • Class I, Division 1 & 2 (Zone 1); Class II, Div. 1 and 2; Class III
  • Worldwide approvals and certifications
  • Terminal block wiring
  • Single conduit
  • Enclosure design permits safe and easy access for wiring
  • External adjustment via reference dial
  • Die cast aluminum enclosure, epoxy powder coated and gasketed