This item is now discontinued. Please browse similar items
United Electric E100-5BS Nema 4x Temperature Switch, -20/80F; Remote Mount
Representative image only.
United Electric Mfr # E100-5BS – Item # RK-39008-21
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • RelaysSPDT
  • Min Temperature (° F)-20
  • Max Temperature (° F)80
  • DescriptionNema 4x Temperature Switch, -20/80F; Remote Mount
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Legendary UE reliability
  • Wideselection of temperature ranges
  • Internal reference scales and adjustment lock
  • Enclosure design permits safe and easy access for wiring
  • Choice of local or remote mount
  • NEMA 4X die cast aluminum enclosure, epoxy powder coated, gasketed, captive screws