This item is now discontinued. Please browse similar items
Troemner WA206 Steel Rubber Coated Lifter for 20kg, 25kg and 30kg
Representative image only.
Troemner Mfr # WA206 – Item # RK-01027-99
SPECS & DESCRIPTION
Specifications & Description
  • Mass CertificationNot applicable
  • DescriptionSteel Rubber Coated Lifter for 20kg, 25kg and 30kg
More About this Item
We offers a variety of lifters and forks of various materials to suit your applications.
Plastic weight forks are constructed of high quality polycarbonate and are designed specific to the size and style of the weight.
Aluminum weight forks and lifters are constructed with an aluminum handle and a PVC
lifting base.
Steel lifters with polyamide coating are designed for ergonomic lifting of large weights.