This item is now discontinued. Please browse similar items
Troemner CLASS 7 5 mg, Class 7 Economical Aluminum Flat Weight with Traceable Certificate
Representative image only.
Troemner Mfr # 61050ST – Item # RK-01028-67
SPECS & DESCRIPTION
Specifications & Description
  • Mass ClassClass 7
  • Mass Size (mg)5
  • Mass CertificationNIST
  • Height (in)0.1406
  • Width (in)0.0625
  • Length (in)0.1406
  • Description5 mg, Class 7 Economical Aluminum Flat Weight with Traceable Certificate
More About this Item
Economical Stainless Steel Weights are economical, quality weights. Individual weights are available from 1 mg through 500 g. Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.
Economical Class 7 weights 500 mg and lower are made of stainless steel or aluminum and are flat, one-piece construction with one side turned up to make them easy to handle with forceps. 500 mg to 100 mg weights are made of stainless steel while weights 100 mg and below are made of aluminum.
Suggested Markets for Economical Stainless Steel Weights include, but are not limited to, educational markets