This item is now discontinued. Please browse similar items
TrippNT 51007BLUE 51007, Rolling Organizer Cart, Blue
Representative image only.
TrippNT Mfr # 51007BLUE – Item # RK-63526-91
Integral handles for easy maneuvering
  • Extra handles for easy maneuvering
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene
  • Description51007, Rolling Organizer Cart, Blue
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Extra handles for easy maneuvering
  • Limited Lifetime Warranty
More About this Item
This rolling cart is loaded with storage space—perfect for organizing and transporting all your lab supplies! Plus, handle cutouts in the top working surface make it easy to maneuver around corners.
Constructed of rustproof, chemical-resistant, high-density polyethylene (HDPE) and ABS plastic, the cart is quickly wiped clean. Sturdy design carries up to three times its weight in supplies and the nonmarking rubber casters roll smoothly on most surfaces. Easy to assemble.
Three full-extension drawers provide full access to small items and stored supplies. Each drawer measures 11-1/2”W x 3-1/2”H x 15-1/2”D (29.2 x 8.9 x 39.4 cm). The large open bulk area measures 12-3/4”W x 15-3/4”H x 16-1/2”D (32.4 x 40 x 41.9 cm).
Use the sliding acrylic door to securely lock either the drawers or the bulk area. Six side shelves hold boxes of gloves, bottles, flasks, and much more.