This item is now discontinued. View the direct replacement: RK-06760-58 or browse similar items
TriForest WPC0400 Wide-Mouth Bottle, 400 mL, PC, Screw, pack of 12, 72/CS
Representative image only.
TriForest Mfr # WPC0400 – Item # RK-06542-33
Engineered for ultra-centrifugation and culturing
  • Culture microbes, plants, or algae—large surface area to volume ratio
  • Polycarbonate (PC) is a safer, shatterproof, alternative to glass
  • Leakproof seals (included) secure samples and prevent aerosol escape
  • Bottle and cap will both withstand reuse and multiple autoclave cycles
  • Molded-in graduations make reference fills easy
  • 12 bottles per pack, 6 packs per case, 72 bottles total
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)400
  • MaterialPolycarbonate(PC)
  • DescriptionTriForest WPC0400 Wide-Mouth Bottle, PC, 400 mL; 72/Cs
Key Features
  • Culture microbes, plants, or algae—large surface area to volume ratio
  • Polycarbonate (PC) is a safer, shatterproof, alternative to glass
  • Leakproof seals (included) secure samples and prevent aerosol escape
  • Bottle and cap will both withstand reuse and multiple autoclave cycles
  • Molded-in graduations make reference fills easy
  • 12 bottles per pack, 6 packs per case, 72 bottles total
More About this Item
Use these clear, shatterproof, polycarbonate bottles to store critical fluids or as ultracentrifuge flasks—bottles withstand maximum speeds without leakage.
Autoclavable