This item is now discontinued. View the direct replacement: RK-06760-56 or browse similar items
TriForest WPC0180 Wide-Mouth Bottle, 180 mL, PC, Screw, pack of 12, 144/CS
Representative image only.
TriForest Mfr # WPC0180 – Item # RK-06542-31
Engineered for ultra-centrifugation and culturing
  • Culture microbes, plants, or algae—large surface area to volume ratio
  • Polycarbonate (PC) is a safer, shatterproof, alternative to glass
  • Leakproof seals (included) secure samples and prevent aerosol escape
  • Bottle and cap will both withstand reuse and multiple autoclave cycles
  • Molded-in graduations make reference fills easy
  • 12 bottles per pack, 12 packs per case, 144 bottles total
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)180
  • MaterialPolycarbonate(PC)
  • DescriptionTriForest WPC0180 Wide-Mouth Bottle, PC, 180 mL; 144/Cs
Key Features
  • Culture microbes, plants, or algae—large surface area to volume ratio
  • Polycarbonate (PC) is a safer, shatterproof, alternative to glass
  • Leakproof seals (included) secure samples and prevent aerosol escape
  • Bottle and cap will both withstand reuse and multiple autoclave cycles
  • Molded-in graduations make reference fills easy
  • 12 bottles per pack, 12 packs per case, 144 bottles total
More About this Item
Use these clear, shatterproof, polycarbonate bottles to store critical fluids or as ultracentrifuge flasks—bottles withstand maximum speeds without leakage.
Autoclavable