This item is now discontinued. Please browse similar items

TriForest Serum Vials, Sterile, 10 mL, PETG, 1600/CS

Representative image only.
TriForest Mfr # RGC0010S – Item # RK-06019-34
Protect your samples with leakproof vials
  • Ideal for safe storage, transport, and transfer of serums and substrates
  • Constructed of optically clear shatterproof plastic

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)10
  • Tube MaterialPETG
  • Tube ColorTransparent
  • Cap TypeTamper Evident Cap
  • Cap Diameter (mm)26.5
  • Cap MaterialPolyethylene
  • Cap colorBlue
  • Thread20-415
  • SterileYes
  • AutoclavableNo
  • Height (in)2 1/2
  • Outside Diameter (in)59/64
  • Height (mm)64.1
  • Outside Diameter (mm)23.7
  • DescriptionSerum Vials, Sterile, 10 mL, PETG, 1920/CS

Key Features

  • Ideal for safe storage, transport, and transfer of serums and substrates
  • Constructed of optically clear shatterproof plastic

More About this Item

The polyethylene terephthalate copolyester (PETG) serum vials are presterilized via gamma irradiation, sterile to SAL 10-6. These vials meet current biological and pharmaceutical regulatory standards including USP Class VI, noncytotoxic, nonpyrogenic, USP 661, European Pharmacopeia for Abnormal toxicity, and are Animal Derived Component Free (ADCF). The PETG vials are cleanroom production packaged with a blue tamper-evident polyethylene (PE) cap engaged. The transparent PETG vials feature molded graduations for reference measurements.
The polycarbonate (PC) serum vials and polypropylene (PP) caps come nonsterile with lot number traceability. These can be autoclaved, up to 125°C, and up to 10 autoclave cycles. The temperature range of these vials is -193 to 257°F (-125 to 125°C). The caps are packaged separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat