This item is now discontinued. Please browse similar items

TriForest Erlenmeyer Flask Clamp, PC, for 500 mL Shaker, each

Representative image only.
TriForest Mfr # SFC0500 – Item # RK-06019-39
Universal replacement clamps for shaking incubators
  • Compatible with all industry-standard shaking incubators
  • Constructed of blue nonscratch polypropylene
  • Autoclavable at 121°C (249°F)

SPECS & DESCRIPTION

Specifications & Description

  • Clamp MaterialPolypropylene
  • Clamp TypeErlenmeyer
  • ColorBlue
  • Height (in)2.5
  • Diameter (in)4.4
  • DescriptionErlenmeyer Flask Clamp, PC, for 500 mL Shaker

Key Features

  • Compatible with all industry-standard shaking incubators
  • Constructed of blue nonscratch polypropylene
  • Autoclavable at 121°C (249°F)

More About this Item

These clamps are suitable for standard shaking incubators and accept Erlenmeyer flasks. Screw the clamp on the platform and snap the flask into the clamp.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat