This item is now discontinued. Please browse similar items

TriForest BPC0030 Square Autoclavable Media Bottle, 30 mL, PC, 24 per pack, 288/CS

Representative image only.
TriForest Mfr # BPC0030 – Item # RK-06542-19
Withstand multiple autoclave cycles
  • Polycarbonate (PC) is a safe and shatterproof alternative to glass
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom in an ISO 9001 compliant facility
  • Keep samples secure—leakproof seals included
  • Easily complete fill with molded-in graduations
  • 24 bottles per pack, 12 packs per case; 288 bottles total

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)30
  • Capacity (oz)1
  • MaterialPolycarbonate(PC)
  • Min Graduation (mL)10
  • Max Graduation (mL)30
  • Graduation Interval (mL)10
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size20/415
  • Cap ColorBlue
  • Bottle ColorTransparent
  • AutoclavableYes
  • Special FeaturesPacked in bags
  • DescriptionSquare/Octagonal Media Bottle, 30 mL; 288/Cs
ISO9001 Certified Supplier

Key Features

  • Polycarbonate (PC) is a safe and shatterproof alternative to glass
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom in an ISO 9001 compliant facility
  • Keep samples secure—leakproof seals included
  • Easily complete fill with molded-in graduations
  • 24 bottles per pack, 12 packs per case; 288 bottles total

More About this Item

Store buffers, reagents, media, and other critical fluids in these sturdy, clear bottles. The square shape saves shelf space by allowing bottles to be placed closer together.

Bottles come separated into packs by bags, and a case is double bagged to maintain cleanliness.

Autoclavable

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat