DescriptionTraceable® Remote-Probe Thermohygrometer with Dew Point and Calibration
Warranty1 year
Key Features
Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Traceable® model number 4189
1 year warranty
More About this Item
Unit simultaneously displays humidity or dew point and temperature. One of the newest and fastest humidity sensors ever developed displays full-range humidity changes in 5-30 seconds. It records wide humidity variations on a second-to-second basis. Monitor labs, clean rooms, food processing, and all humidity-sensitive manufacturing areas with this high-resolution unit. Relative humidity resolution is 0.01%, accuracy is ±3% RH, and range is 10.00 to 95.00% RH. Dew Point is a calculated measurement, resolution is 0.01°, accuracy is sum accuracy of humidity and temperature. Range is –13.50 to 120.1°F and –25.30 to 48.90°C. Temperature resolution is 0.01°, accuracy is ±1°C, and range is 32.00 to 122.0°F and 0.00 to 50.00°C. Memory recalls minimum/maximum readings. HOLD button freezes display for reading later. Data logger captures 1000 readings manually at the push of a button or automatically when set between 1 to 3600 seconds. Computer output allows unit to be connected to a computer for down loading data logger results or constant monitoring. Jumbo-size display is 1-3/8-inch-high. Case is impact-resistant ABS plastic. Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.