Traceable Calibrated Compact Two-Channel Digital Timer

Representative image only.
Traceable Item # RK-94461-31
Stream-lined timer can be carried and place anywhere
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

$28.36CAD / Each

BUY MORE & SAVE

  • 1-9$28.36CAD / Each
  • 10+$25.52CAD / Each
Time to recalibrate?
for an additional
$98.42CAD / Each
for an additional
$205.54CAD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Timing Capacity24 hours
  • Count DirectionUp and down
  • ClockYes
  • Number of Channels2
  • Resolution1 sec
  • Accuracy±0.01%
  • AlarmDistinct alarm for each channel
  • DisplayLCD, 3/8\" H digits
  • CertificationsNIST-traceable
  • Depth (in)2
  • Width (in)2 1/2
  • Depth (cm)5.1
  • Width (cm)6.4
  • DescriptionCalibrated Compact Two-Channel Digital Timer
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration

Key Features

  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

Compact size and two channels make this timer ideal for the lab. Set two channels independently in count-up or countdown mode. Distinctive alarm sound for each channel beeps for one minute after set time has elapsed. For repetitive tests, memory function automatically returns the display to the original set time.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.

Guaranteed to have at least one year of calibration remaining on certificate.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat