Total Source APP0380-21-20 Polyethylene (PE)-Coated Polypropylene (PP) Sleeves, 20" Length, Case of 200 (100 Pairs)
Representative image only.
Total Source Mfr # APP0380-21-20 – Item # RK-86227-72
Splash protection in a comfortable, breathable material
  • Ideal for general-purpose laboratory use
PRODUCT OPTIONS
$146.45CAD / Case of 200
BUY MORE & SAVE
  • 1-9$146.45CAD / Case of 200
  • 10+$131.81CAD / Case of 200
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)20
  • Length (cm)50.8
  • MaterialPolyethylene(PE)
  • ColorWhite
  • DescriptionPolyethylene (PE)-Coated Polypropylene (PP) Sleeves, 20\" Length
Key Features
  • Ideal for general-purpose laboratory use
More About this Item
These polyethylene-coated polypropylene sleeves are a low cost, disposable alternative to Tyvek®. The fabric is durable, lightweight, and breathable. The PE-coated spun bond PP provides excellent splash protection from water based liquids.