Total Source APP0242-W Polyethylene-Coated Polypropylene Lab Coats, Large, case of 30
Representative image only.
Total Source Mfr # APP0242-W-L – Item # RK-86227-14
Excellent lightweight splash protection
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
PRODUCT OPTIONS
$202.22CAD / Case of 30
BUY MORE & SAVE
  • 1-9$202.22CAD / Case of 30
  • 10+$182.00CAD / Case of 30
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)41.5
  • Length (cm)105.41
  • SizeLarge
  • MaterialPolyethylene(PE)
  • ColorWhite
  • DescriptionPolyethylene-Coated Polypropylene Lab Coats, Large
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
These polyethylene (PE) coated polypropylene (PP) lab coats are a low-cost disposable alternative to Tyvek® coats. The durable, lightweight, and breathable fabric is ideal for general-purpose laboratory applications. The PE-coated spun bond PP-provides excellent splash protection from water-based liquids.