This item is now discontinued. Please browse similar items
Todd Systems SU-13 G Step-Up Transformer, 1000 Watts
Representative image only.
Todd Systems Mfr # SU-13 G – Item # RK-01578-18
Power your instruments with 110/120 or 220/240 VAC outlets
  • Includes rugged NEMA cord with three-prong plug
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Input (VAC)110to120
  • Output (VAC)220/240
  • Power Rating (watts)1000
  • DescriptionStep-Up Transformer, 1000 Watts
  • Warranty1 Year
1 Year Warranty
Key Features
  • Includes rugged NEMA cord with three-prong plug
  • 1 year warranty
More About this Item
Select a step-down transformer to connect 110/120 VAC instruments to 220/240 VAC power sources; use a step-up transformer to connect 220/240 VAC instruments to 110/120 VAC power sources. All models run on 50/60 Hz.