This item is now discontinued. Please browse similar items

ThermoSafe 334 Multipurpose Mailers, 50/cs

Representative image only.
ThermoSafe Mfr # 334 – Item # RK-03741-30
Ship non-hazardous specimens safely
  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

SPECS & DESCRIPTION

Specifications & Description

  • Inside Depth (in)1 1/2
  • Inside Length (in)8
  • Inside Width (in)2 1/4
  • Outside Width (in)3
  • Outside Length (in)9 1/2
  • Outside Depth (in)2 3/4
  • Inside Depth (cm)3.81
  • Inside Length (cm)20.3
  • Outside Width (cm)7.6
  • Outside Length (cm)24.1
  • Outside Depth (cm)7
  • Shipping Weight (oz)72
  • Shipping Weight (g)2041
  • DescriptionMultipurpose Mailers, 50/cs

Key Features

  • Fitted halves cushion tubes for superior protection
  • Lightweight mailers meet postal regulations for nonhazardous materials

More About this Item

Protect your fragile samples with this expanded polystyrene shipper. Light weight minimizes postage costs. Strong white paperboard shipping carton (sold separately) has sure-lock tabs that eliminate taping or tying. Choose from tube mailers, slide mailer, mini mailer, multipurpose mailer, or aluminum shipping can.

Holds cans, screw-cap vials, tubes, swabs, and air filter cartridges.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat