This item is now discontinued. Please browse similar items
Thermo Scientific VersaCool VersaRack , Adjustable SS Bath Rack
Representative image only.
Thermo Scientific VersaCool Mfr # 230000001 – Item # RK-12128-85
Optimize performance of your Thermo Scientific VersaCool baths
  • Keep your bath running properly with these accessories
  • Utilize the VersaRack for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
  • Holds up to 75 test tubes with 10 mm diameter
  • Holds up to 48 test tubes with 16 mm diameter
  • Holds up to 21 test tubes with 25 mm diameter
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsSS
  • Description, Adjustable SS Bath Rack
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your bath running properly with these accessories
  • Utilize the VersaRack for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
  • Holds up to 75 test tubes with 10 mm diameter
  • Holds up to 48 test tubes with 16 mm diameter
  • Holds up to 21 test tubes with 25 mm diameter
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your VersaCool baths.
Stainless steel adjustable rack for VersaCool bath also acts as a shelf when using beakers and flasks. No tools are needed to adjust rack height or spacing for holding various test tube sizes.