This item is now discontinued. Please browse similar items
Thermo Scientific VersaCool 230000006 Plumbing Adapters Kit
Representative image only.
Thermo Scientific VersaCool Mfr # 230000006 – Item # RK-12128-89
Optimize performance of your Thermo Scientific VersaCool baths
  • Keep your bath running properly with these accessories
  • Replacement stainless steel hose barb and threaded adapter kit
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialsSS
  • DescriptionPlumbing Adapters Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your bath running properly with these accessories
  • Replacement stainless steel hose barb and threaded adapter kit
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your VersaCool baths.
A complete selection of the suggested and common hose barb and threaded adapters used with the VersaCool bath.
Includes 1/4" (6-mm), 8-mm, and 1/2" (12-mm) hosebarb adapters, as well as 1/4" and 1/2" male NPT threaded adapters — perfect for coupling with Quick-Disconnect fittings!